siRNA Details
Gene Symbol: Hccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191732.1 | NP_001178661.1 | 7 | 936 | ||
XM_006256834.2 | XP_006256896.1 | 7 | 910 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191732.1 | NP_001178661.1 | 7 | 936 | ||
XM_006256834.2 | XP_006256896.1 | 7 | 910 |