siRNA Details
Gene Symbol: Baz2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108260.1 | NP_001101730.1 | 36 | 6458 | ||
XM_008761847.1 | XP_008760069.1 | 34 | 6016 | |||
XM_008761848.1 | XP_008760070.1 | 33 | 5974 | |||
XM_008761849.1 | XP_008760071.1 | 34 | 6010 | |||
XM_008761850.1 | XP_008760072.1 | 33 | 5983 | |||
XM_008761851.1 | XP_008760073.1 | 34 | 5911 | |||
XM_008761852.1 | XP_008760074.1 | 34 | 5899 | |||
XM_008761853.1 | XP_008760075.1 | 33 | 5878 | |||
XM_008761854.1 | XP_008760076.1 | 33 | 5872 | |||
XM_008761855.1 | XP_008760077.1 | 33 | 5827 | |||
XM_008761856.1 | XP_008760078.1 | 32 | 5785 | |||
XM_008761857.1 | XP_008760079.1 | 33 | 5821 | |||
XM_008761858.1 | XP_008760080.1 | 33 | 5728 |
