siRNA Details
| Gene Symbol: Bloc1s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025714.1 | NP_001020885.1 | 2 | 266 | ||
| GenBank mRNA | BC097469.1 | - | 2 | 266 | 7457729 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bloc1s6| Gene Symbol: Bloc1s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025714.1 | NP_001020885.1 | 2 | 266 | ||
| GenBank mRNA | BC097469.1 | - | 2 | 266 | 7457729 | |