siRNA Details
| Gene Symbol: Hnrnpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175603.2 | NP_783193.2 | 3 | 270 | ||
| GenBank mRNA | AY184814.1 | - | 2 | 184 | ||
| BC093598.1 | - | 3 | 270 | 7315896 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnpr| Gene Symbol: Hnrnpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175603.2 | NP_783193.2 | 3 | 270 | ||
| GenBank mRNA | AY184814.1 | - | 2 | 184 | ||
| BC093598.1 | - | 3 | 270 | 7315896 | ||