siRNA Details
| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175604.2 | NP_783194.2 | 2 | 493 | ||
| GenBank mRNA | AY172974.1 | - | 219 | |||
| BC088428.1 | - | 2 | 458 | |||
| BC099797.1 | - | 2 | 493 | 7936110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yrdc| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175604.2 | NP_783194.2 | 2 | 493 | ||
| GenBank mRNA | AY172974.1 | - | 219 | |||
| BC088428.1 | - | 2 | 458 | |||
| BC099797.1 | - | 2 | 493 | 7936110 | ||