siRNA Details
| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175604.2 | NP_783194.2 | 4 | 745 | ||
| GenBank mRNA | BC088428.1 | - | 4 | 710 | ||
| BC099797.1 | - | 4 | 745 | 7936110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yrdc| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175604.2 | NP_783194.2 | 4 | 745 | ||
| GenBank mRNA | BC088428.1 | - | 4 | 710 | ||
| BC099797.1 | - | 4 | 745 | 7936110 | ||