siRNA Details
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182825.1 | NP_877977.1 | 7, 8 | 652 | ||
GenBank mRNA | AY327411.1 | - | 7, 8 | 652 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182825.1 | NP_877977.1 | 7, 8 | 652 | ||
GenBank mRNA | AY327411.1 | - | 7, 8 | 652 |