siRNA Details
| Gene Symbol: Asb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236158.2 | XP_006236220.1 | 9 | 1858 | ||
| XM_008762831.1 | XP_008761053.1 | 9 | 2014 | |||
| XM_008762832.1 | XP_008761054.1 | 9 | 1815 | |||
| XM_008762833.1 | XP_008761055.1 | 9 | 2036 | |||

Asb15
Genomic Map