siRNA Details
| Gene Symbol: Figf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031761.1 | NP_113949.1 | 6 | 1058 | ||
| XM_006256863.2 | XP_006256925.1 | 7 | 1205 | |||
| XM_006256864.2 | XP_006256926.1 | 6 | 1535 | |||
| GenBank mRNA | AF014827.1 | - | 6 | 1058 | ||
| AY032728.1 | - | 6 | 833 | |||

Figf
Genomic Map