siRNA Details
| Gene Symbol: Figf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031761.1 | NP_113949.1 | 2 | 428 | ||
| GenBank mRNA | AF014827.1 | - | 2 | 428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Figf| Gene Symbol: Figf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031761.1 | NP_113949.1 | 2 | 428 | ||
| GenBank mRNA | AF014827.1 | - | 2 | 428 | ||