siRNA Details
| Gene Symbol: Tfap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108267.1 | NP_001101737.1 | 4 | 711 | ||
| XM_006245818.2 | XP_006245880.1 | 4 | 1467 | |||
| XM_006245819.2 | XP_006245881.1 | 4 | 1166 | |||
| XM_006245820.2 | XP_006245882.1 | 4 | 1603 | |||
| XM_006245822.2 | XP_006245884.1 | 4 | 2293 | |||
| XM_006245823.2 | XP_006245885.1 | 4 | 1127 | |||

Tfap4
Genomic Map