siRNA Details
| Gene Symbol: Jmjd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014116.1 | NP_001014138.1 | 8 | 853 | ||
| XM_006245998.2 | XP_006246060.1 | 7 | 947 | |||
| XM_006245999.2 | XP_006246061.1 | 8 | 866 | |||
| GenBank mRNA | BC079205.1 | - | 8 | 853 | 7131778 | |

Jmjd8
Genomic Map