siRNA Details
| Gene Symbol: Dock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767630.1 | XP_008765852.1 | 21 | 2179 | ||
| GenBank mRNA | BC101877.1 | - | 21 | 2134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock2| Gene Symbol: Dock2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767630.1 | XP_008765852.1 | 21 | 2179 | ||
| GenBank mRNA | BC101877.1 | - | 21 | 2134 | ||