siRNA Details
| Gene Symbol: Zcchc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271026.1 | NP_001257955.1 | 2, 3 | 206 | ||
| GenBank mRNA | AY387053.1 | - | 652 | |||
| BC089894.1 | - | 2, 3 | 206 | |||
| BC158679.1 | - | 2, 3 | 206 | |||
| FQ227271.1 | - | 2, 3 | 227 | |||

Zcchc10
Genomic Map