siRNA Details
| Gene Symbol: Slc47a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014118.2 | NP_001014140.1 | 17 | 1496 | ||
| GenBank mRNA | AB248823.1 | - | 17 | 1496 | ||
| AB248824.1 | - | 17 | 1496 | |||
| BC088413.1 | - | 1516 | 7109897 | |||
| FQ217802.1 | - | 1510 | ||||

Slc47a1
Genomic Map