siRNA Details
| Gene Symbol: Phf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004272.1 | NP_001004272.1 | 4 | 377 | ||
| XM_006246747.2 | XP_006246809.1 | 4 | 634 | |||
| XM_006246748.1 | XP_006246810.1 | 4 | 529 | |||
| XM_008767840.1 | XP_008766062.1 | 4 | 328 | |||
| GenBank mRNA | BC079162.1 | - | 4 | 377 | 7129493 | |

Phf23
Genomic Map