siRNA Details
| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024270.2 | NP_001019441.2 | 9, 10 | 1073 | ||
| GenBank mRNA | AY970831.2 | - | 9, 10 | 1071 | ||
| BC101890.1 | - | 9, 10 | 1073 | 7930679 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pelp1| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024270.2 | NP_001019441.2 | 9, 10 | 1073 | ||
| GenBank mRNA | AY970831.2 | - | 9, 10 | 1071 | ||
| BC101890.1 | - | 9, 10 | 1073 | 7930679 | ||