siRNA Details
| Gene Symbol: Utp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768149.1 | XP_008766371.1 | 19 | 1866 | ||
| GenBank mRNA | FQ210246.1 | - | 19 | 1834 | ||
| FQ217168.1 | - | 19 | 1830 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Utp6| Gene Symbol: Utp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768149.1 | XP_008766371.1 | 19 | 1866 | ||
| GenBank mRNA | FQ210246.1 | - | 19 | 1834 | ||
| FQ217168.1 | - | 19 | 1830 | |||