siRNA Details
Gene Symbol: Ints2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100630.2 | NP_001094100.2 | 11 | 1800 | ||
XM_008768072.1 | XP_008766294.1 | 11 | 1867 | |||
GenBank mRNA | BC128757.1 | - | 11 | 1773 | 7107619 |
