siRNA Details
| Gene Symbol: Mrps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108289.1 | NP_001101759.1 | 4 | 406 | ||
| XM_008768075.1 | XP_008766297.1 | 4 | 597 | |||
| GenBank mRNA | FQ213501.1 | - | 4 | 942 | ||
| FQ216939.1 | - | 4 | 437 | |||

Mrps23
Genomic Map