siRNA Details
| Gene Symbol: Igfbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004274.2 | NP_001004274.1 | 3, 4 | 875 | ||
| GenBank mRNA | AY686592.1 | - | 3, 4 | 639 | ||
| BC098750.1 | - | 3, 4 | 875 | 7368649 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp4| Gene Symbol: Igfbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004274.2 | NP_001004274.1 | 3, 4 | 875 | ||
| GenBank mRNA | AY686592.1 | - | 3, 4 | 639 | ||
| BC098750.1 | - | 3, 4 | 875 | 7368649 | ||