siRNA Details
| Gene Symbol: Etv4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108299.1 | NP_001101769.1 | 11 | 1385 | ||
| XM_006247369.2 | XP_006247431.1 | 11 | 1458 | |||
| XM_006247370.2 | XP_006247432.1 | 11 | 1406 | |||
| XM_006247371.2 | XP_006247433.1 | 11 | 1366 | |||
| XM_006247372.2 | XP_006247434.1 | 11 | 1213 | |||
| XM_008768085.1 | XP_008766307.1 | 11 | 1455 | |||
| XM_008768086.1 | XP_008766308.1 | 11 | 1441 | |||
| GenBank mRNA | BC168950.1 | - | 11 | 1259 | 9022277 | |

Etv4
Genomic Map