siRNA Details
Gene Symbol: Taco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108302.1 | NP_001101772.1 | 5 | 1063 | ||
GenBank mRNA | BC166900.1 | - | 5 | 1014 | 6922142 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Taco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108302.1 | NP_001101772.1 | 5 | 1063 | ||
GenBank mRNA | BC166900.1 | - | 5 | 1014 | 6922142 |