siRNA Details
| Gene Symbol: Gga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108304.1 | NP_001101774.1 | 10 | 1987 | ||
| XM_006247719.2 | XP_006247781.1 | 10 | 966 | |||
| XM_006247720.2 | XP_006247782.1 | 8 | 1104 | |||
| XM_006247721.1 | XP_006247783.1 | 9 | 790 | |||

Gga3
Genomic Map