siRNA Details
Gene Symbol: Dscr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108316.2 | NP_001101786.2 | 7, 8 | 824 | ||
XM_006248072.2 | XP_006248134.1 | 6, 7 | 1128 | |||
XM_006248073.2 | XP_006248135.1 | 6, 7 | 1126 | |||
XM_008768583.1 | XP_008766805.1 | 7, 8 | 1212 | |||
GenBank mRNA | BC160826.1 | - | 7, 8 | 824 | 7444837 |
