siRNA Details
| Gene Symbol: Ccl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037203.1 | NP_001032280.1 | 3 | 245 | ||
| XM_006248766.2 | XP_006248828.1 | 3 | 355 | |||
| GenBank mRNA | BC107945.1 | - | 3 | 245 | 7388091 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl25| Gene Symbol: Ccl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037203.1 | NP_001032280.1 | 3 | 245 | ||
| XM_006248766.2 | XP_006248828.1 | 3 | 355 | |||
| GenBank mRNA | BC107945.1 | - | 3 | 245 | 7388091 | |