siRNA Details
| Gene Symbol: Rhbdd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191827.1 | NP_001178756.1 | 2 | 339 | ||
| XM_006249182.2 | XP_006249244.1 | 2 | 331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdd2| Gene Symbol: Rhbdd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191827.1 | NP_001178756.1 | 2 | 339 | ||
| XM_006249182.2 | XP_006249244.1 | 2 | 331 | |||