siRNA Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012147.1 | NP_001012147.1 | 3 | 456 | ||
| XM_006249455.2 | XP_006249517.1 | 3 | 462 | |||
| XM_006249457.2 | XP_006249519.1 | 3 | 1206 | |||
| XM_006249459.2 | XP_006249521.1 | 3 | 1205 | |||
| XM_006249460.2 | XP_006249522.1 | 3 | 462 | |||
| GenBank mRNA | AY641810.1 | - | 3 | 457 | ||
| BC081984.1 | - | 3 | 456 | 7128801 | ||
| FQ226157.1 | - | 3 | 458 | |||

Pxn
Genomic Map