siRNA Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108341.1 | NP_001101811.1 | 16 | 1384 | ||
| XM_006249565.2 | XP_006249627.1 | 16 | 1690 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ulk1| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108341.1 | NP_001101811.1 | 16 | 1384 | ||
| XM_006249565.2 | XP_006249627.1 | 16 | 1690 | |||