siRNA Details
| Gene Symbol: Tada1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037980.2 | NP_001033069.2 | 5 | 503 | ||
| GenBank mRNA | BC091380.1 | - | 295 | 7320760 | ||
| BC158713.1 | - | 5 | 503 | 9035442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tada1| Gene Symbol: Tada1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037980.2 | NP_001033069.2 | 5 | 503 | ||
| GenBank mRNA | BC091380.1 | - | 295 | 7320760 | ||
| BC158713.1 | - | 5 | 503 | 9035442 | ||