siRNA Details
| Gene Symbol: Taf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037204.1 | NP_001032281.1 | 9 | 1125 | ||
| XM_008769862.1 | XP_008768084.1 | 9 | 1147 | |||
| GenBank mRNA | BC107466.1 | - | 9 | 1125 | 7460374 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taf1a| Gene Symbol: Taf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037204.1 | NP_001032281.1 | 9 | 1125 | ||
| XM_008769862.1 | XP_008768084.1 | 9 | 1147 | |||
| GenBank mRNA | BC107466.1 | - | 9 | 1125 | 7460374 | |