siRNA Details
Gene Symbol: Cnot6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108355.1 | NP_001101825.1 | 9 | 1041 | ||
XM_006250708.2 | XP_006250770.1 | 9 | 1073 | |||
XM_006250709.2 | XP_006250771.1 | 9 | 960 | |||
XM_008770042.1 | XP_008768264.1 | 8 | 3840 | |||
XM_008770043.1 | XP_008768265.1 | 8 | 852 |
