siRNA Details
| Gene Symbol: Stk24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127494.1 | NP_001120966.1 | 253 | |||
| GenBank mRNA | EU371958.1 | - | 253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk24| Gene Symbol: Stk24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127494.1 | NP_001120966.1 | 253 | |||
| GenBank mRNA | EU371958.1 | - | 253 | |||