siRNA Details
| Gene Symbol: Galntl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135756.1 | NP_001129228.1 | 12 | 1904 | ||
| XM_008771227.1 | XP_008769449.1 | 11 | 2558 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galntl6| Gene Symbol: Galntl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135756.1 | NP_001129228.1 | 12 | 1904 | ||
| XM_008771227.1 | XP_008769449.1 | 11 | 2558 | |||