siRNA Details
Gene Symbol: Rab24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015023.1 | NP_001015023.1 | 3 | 641 | ||
XM_006253632.2 | XP_006253694.1 | 3 | 532 | |||
GenBank mRNA | BC091184.1 | - | 3 | 641 | 7366064 |
