siRNA Details
Gene Symbol: Rab24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015023.1 | NP_001015023.1 | 5 | 851 | ||
GenBank mRNA | BC091184.1 | - | 5 | 851 | 7366064 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015023.1 | NP_001015023.1 | 5 | 851 | ||
GenBank mRNA | BC091184.1 | - | 5 | 851 | 7366064 |