siRNA Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170343.1 | NP_001163814.1 | 12 | 1419 | ||
| XM_006254257.2 | XP_006254319.1 | 10 | 1255 | |||
| GenBank mRNA | CB327956.1 | - | 1 | 619 | ||
| CO567453.1 | - | 595 | ||||

Sec61a2
Genomic Map