siRNA Details
| Gene Symbol: Nudt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007733.1 | NP_001007734.1 | 4, 5 | 329 | ||
| XM_008771875.1 | XP_008770097.1 | 4, 5 | 348 | |||
| GenBank mRNA | BC079176.1 | - | 4, 5 | 329 | 7130276 | |
| FQ219814.1 | - | 2, 3 | 72 | |||
| FQ228004.1 | - | 2, 3 | 72 | |||

Nudt5
Genomic Map