siRNA Details
| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108440.1 | NP_001101910.1 | 14, 15 | 1488 | ||
| XM_006255267.2 | XP_006255329.1 | 15, 16 | 2023 | |||
| XM_006255268.2 | XP_006255330.1 | 15, 16 | 2020 | |||
| XM_008772409.1 | XP_008770631.1 | 12, 13 | 1629 | |||

Brd7
Genomic Map