siRNA Details
| Gene Symbol: Dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108441.1 | NP_001101911.1 | 2 | 714 | ||
| XM_006255282.2 | XP_006255344.1 | 2 | 681 | |||
| GenBank mRNA | BC159430.1 | - | 2 | 577 | 9089687 | |
| FQ227962.1 | - | 2 | 635 | |||

Dnajb1
Genomic Map