siRNA Details
Gene Symbol: Gab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108444.1 | NP_001101914.1 | 10 | 2284 | ||
GenBank mRNA | GQ140625.1 | - | 11 | 2084 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108444.1 | NP_001101914.1 | 10 | 2284 | ||
GenBank mRNA | GQ140625.1 | - | 11 | 2084 |