siRNA Details
| Gene Symbol: Prmt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014153.1 | NP_001014175.1 | 10 | 1259 | ||
| GenBank mRNA | BC085121.1 | - | 10 | 1259 | 7115238 | |
| FQ211965.1 | - | 1302 | ||||
| FQ212485.1 | - | 1226 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt7| Gene Symbol: Prmt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014153.1 | NP_001014175.1 | 10 | 1259 | ||
| GenBank mRNA | BC085121.1 | - | 10 | 1259 | 7115238 | |
| FQ211965.1 | - | 1302 | ||||
| FQ212485.1 | - | 1226 | ||||