siRNA Details
| Gene Symbol: Cbfa2t3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108453.1 | NP_001101923.1 | 3, 4 | 384 | ||
| XM_006255772.2 | XP_006255834.1 | 3, 4 | 539 | |||
| XM_008772629.1 | XP_008770851.1 | 3, 4 | 2054 | |||
| XM_008772631.1 | XP_008770853.1 | 3, 4 | 539 | |||

Cbfa2t3
Genomic Map