siRNA Details
| Gene Symbol: Sipa1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009704.1 | NP_001009704.1 | 14 | 4069 | ||
| XM_008772638.1 | XP_008770860.1 | 14 | 4093 | |||
| XM_008772639.1 | XP_008770861.1 | 14 | 4093 | |||
| GenBank mRNA | AY043227.1 | - | 14 | 4069 | ||

Sipa1l2
Genomic Map