siRNA Details
| Gene Symbol: Ntpcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134573.1 | NP_001128045.1 | 4 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntpcr| Gene Symbol: Ntpcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134573.1 | NP_001128045.1 | 4 | 397 | ||