siRNA Details
| Gene Symbol: Ppp6r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135849.1 | NP_001129321.1 | 4 | 1021 | ||
| XM_006228275.2 | XP_006228337.1 | 4 | 1280 | |||
| XM_006228276.2 | XP_006228338.1 | 4 | 1279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp6r1| Gene Symbol: Ppp6r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135849.1 | NP_001129321.1 | 4 | 1021 | ||
| XM_006228275.2 | XP_006228337.1 | 4 | 1280 | |||
| XM_006228276.2 | XP_006228338.1 | 4 | 1279 | |||