siRNA Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012167.1 | NP_001012167.1 | 9 | 1031 | ||
| XM_006228578.2 | XP_006228640.1 | 11 | 1360 | |||
| XM_006228579.2 | XP_006228641.1 | 11 | 1308 | |||
| XM_006228580.2 | XP_006228642.1 | 10 | 1466 | |||
| XM_006228581.2 | XP_006228643.1 | 11 | 1338 | |||
| XM_008759165.1 | XP_008757387.1 | 10 | 1251 | |||
| GenBank mRNA | BC089987.1 | - | 9 | 1031 | 7318612 | |

Pld3
Genomic Map