siRNA Details
| Gene Symbol: Trpm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037733.1 | NP_001032822.1 | 27 | 4634 | ||
| NM_001037734.1 | NP_001032823.1 | 27 | 4652 | |||
| XM_006229344.2 | XP_006229406.1 | 26 | 4582 | |||
| XM_006229345.2 | XP_006229407.1 | 26 | 4579 | |||
| GenBank mRNA | AJ867482.1 | - | 27 | 4652 | ||
| AJ867483.1 | - | 27 | 4634 | |||

Trpm1
Genomic Map