siRNA Details
Gene Symbol: Aamdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108493.1 | NP_001101963.1 | 3 | 360 | ||
XM_006229768.1 | XP_006229830.1 | 2 | 269 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aamdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108493.1 | NP_001101963.1 | 3 | 360 | ||
XM_006229768.1 | XP_006229830.1 | 2 | 269 |