siRNA Details
| Gene Symbol: Arap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229897.2 | XP_006229959.1 | 22 | 3287 | ||
| XM_006229898.2 | XP_006229960.1 | 22 | 4031 | |||
| XM_006229899.2 | XP_006229961.1 | 22 | 4029 | |||
| XM_006229900.1 | XP_006229962.1 | 20 | 2906 | |||
| XM_006229901.1 | XP_006229963.1 | 19 | 2727 | |||
| XM_008759810.1 | XP_008758032.1 | 23 | 4454 | |||
| XM_008759811.1 | XP_008758033.1 | 22 | 4029 | |||

Arap1
Genomic Map